Assay Details
Target Gene Details
Entrez Gene ID: | 3983 |
Gene Name: | actin binding LIM protein 1 |
Gene Aliases: |
ABLIM, LIMAB1, LIMATIN, abLIM-1 |
Location: |
Chr.10:114431110-114821643 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 35 - Exon 36 |
More Information
Set Membership: |
|

Genomic Map