Assay Details
Target Gene Details
Entrez Gene ID: | 57680 |
Gene Name: | chromodomain helicase DNA binding protein 8 |
Gene Aliases: |
AUTS18, HELSNF1 |
Location: |
Chr.14:21385194-21437298 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD8 | NM_001170629.1 | NP_001164100.1 | ||
| NM_020920.3 | NP_065971.2 | |||
| AB046784.3 | BAB13390.3 | |||
| AK131077.1 | BAC85127.1 | |||
| AL834524.1 | CAD39180.1 | |||
| BC011695.2 | AAH11695.2 | |||
| BC025964.1 | AAH25964.1 | |||
| BC036920.1 | AAH36920.1 | |||
| BC063693.1 | AAH63693.1 | |||
| BC073903.1 | AAH73903.1 | |||
| BC098452.1 | AAH98452.1 | |||
| CR749315.1 | CAH18170.1 |
More Information
Set Membership: |
|

Genomic Map