Assay Details
Target Gene Details
Entrez Gene ID: | 51267 |
Gene Name: | C-type lectin domain family 1 member A |
Gene Aliases: |
CLEC-1, CLEC1 |
Location: |
Chr.12:10069504-10099065 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC1A | NM_001297748.1 | 5 | 809 | NP_001284677.1 |
| NM_001297749.1 | 4 | 632 | NP_001284678.1 | |
| NM_001297750.1 | 7 | 1006 | NP_001284679.1 | |
| NM_001297751.1 | 6 | 950 | NP_001284680.1 | |
| NM_016511.3 | 6 | 908 | NP_057595.2 | |
| XM_011520687.2 | 7 | 989 | XP_011518989.1 | |
| AF200949.1 | 6 | 846 | AAF36830.1 | |
| AK075114.1 | 6 | 828 | BAC11410.1 | |
| AK091989.1 | 5 | 760 | ||
| AK291584.1 | 6 | 796 | ||
| AK300979.1 | 4 | 519 | ||
| AK313589.1 | 6 | 826 | ||
| AY358587.1 | 6 | 908 | AAQ88950.1 | |
| BC039072.1 | 6 | 814 | AAH39072.1 | |
| BC067746.1 | 6 | 828 | AAH67746.1 | |
| BQ722745.1 | 6 | 837 | ||
| BX339075.2 | 6 | 846 | ||
| BX380212.2 | 7 | 863 |
More Information
Additional Information:
For this assay, SNP(s) [rs77910243] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map