Assay Details
Target Gene Details
Entrez Gene ID: | 283383 |
Gene Name: | adhesion G protein-coupled receptor D1 |
Gene Aliases: |
GPR133, PGR25 |
Location: |
Chr.12:130953907-131141469 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRD1 | NM_198827.3 | NP_942122.2 | ||
| XM_005253566.2 | XP_005253623.1 | |||
| XM_011538203.2 | XP_011536505.1 | |||
| XM_011538204.2 | XP_011536506.1 | |||
| XM_011538205.2 | XP_011536507.1 | |||
| XM_011538206.2 | XP_011536508.1 | |||
| XM_011538207.2 | XP_011536509.1 | |||
| XM_011538208.2 | XP_011536510.1 | |||
| XM_011538209.2 | XP_011536511.1 | |||
| XM_011538210.2 | XP_011536512.1 | |||
| AY532280.1 | AAS37682.1 | |||
| BC112307.1 | AAI12308.1 | |||
| BC112309.1 | AAI12310.1 | |||
| BC143775.1 | ||||
| BX649116.1 | 3 | 751 | ||
| EU523118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038008 | Chr.12:130928831 - 131167119 on Build GRCh38 | Gain |
|
| nsv560769 | Chr.12:130916107 - 131621821 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map