Assay Details
Target Gene Details
Entrez Gene ID: | 5269 |
Gene Name: | serpin family B member 6 |
Gene Aliases: |
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3 |
Location: |
Chr.6:2948159-2972165 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB6 | NM_001195291.2 | 8 | 2005 | NP_001182220.2 |
| NM_001271822.1 | 7 | 1430 | NP_001258751.1 | |
| NM_001271823.1 | 7 | 1528 | NP_001258752.1 | |
| NM_001271824.1 | 7 | 1912 | NP_001258753.1 | |
| NM_001271825.1 | 7 | 1514 | NP_001258754.1 | |
| NM_001297699.1 | 7 | 2155 | NP_001284628.1 | |
| NM_001297700.1 | 7 | 1811 | NP_001284629.1 | |
| NM_004568.5 | 7 | 1870 | NP_004559.4 | |
| XM_011514672.1 | 9 | 1808 | XP_011512974.1 | |
| XM_011514673.1 | 7 | 1962 | XP_011512975.1 | |
| XM_011514674.2 | 7 | 1547 | XP_011512976.1 | |
| XM_011514676.2 | 7 | 1357 | XP_011512978.1 | |
| XM_017010940.1 | 8 | 1369 | XP_016866429.1 | |
| XM_017010941.1 | 6 | 2998 | XP_016866430.1 | |
| AF116727.1 | AAO15303.1 | |||
| AK027149.1 | 6 | 2696 | ||
| AK057138.1 | 6 | 3229 | ||
| AK094983.1 | ||||
| AK098566.1 | ||||
| AK122895.1 | ||||
| AK122961.1 | 6 | 2146 | ||
| AK128637.1 | ||||
| BC001394.2 | 7 | 1452 | AAH01394.1 | |
| BC098564.1 | 7 | 1308 | AAH98564.1 | |
| BX538343.1 | CAD98106.1 | |||
| Z22658.1 | 7 | 1301 | CAA80373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950760 | Chr.6:2946367 - 2957166 on Build GRCh38 | Deletion |
|
| nsv437477 | Chr.6:2934407 - 2948731 on Build GRCh38 | Loss |
|
| nsv600832 | Chr.6:2945097 - 2970334 on Build GRCh38 | Loss |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
| nsv1018369 | Chr.6:2944928 - 2961123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map