Assay Details
Target Gene Details
Entrez Gene ID: | 102546294 |
Gene Name: | uncharacterized LOC102546294 |
Gene Aliases: |
- |
Location: |
Chr.5:148268307-148383850 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102546294 | NR_105057.1 | |||
| AK054753.1 | ||||
| DA523579.1 |
Target Gene Details
Entrez Gene ID: | 84651 |
Gene Name: | serine peptidase inhibitor, Kazal type 7 (putative) |
Gene Aliases: |
ECG2, ECRG2 |
Location: |
Chr.5:148312427-148315919 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK7 | NM_032566.2 | NP_115955.1 | ||
| AF268198.1 | AAK27795.1 | |||
| AK311570.1 | 2 | 871 | ||
| AY359072.1 | AAQ89431.1 | |||
| BC109385.1 | AAI09386.1 | |||
| BC110067.1 | AAI10068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607102 | Chr.5:148026584 - 148359140 on Build GRCh38 | Loss |
|
| esv3607106 | Chr.5:148224310 - 148363295 on Build GRCh38 | Loss |
|
| nsv5058 | Chr.5:148313602 - 148346355 on Build GRCh38 | Insertion |
|
| nsv599945 | Chr.5:148269618 - 148340924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map