Assay Details
Target Gene Details
Entrez Gene ID: | 80724 |
Gene Name: | acyl-CoA dehydrogenase family member 10 |
Gene Aliases: |
- |
Location: |
Chr.12:111686053-111757107 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAD10 | NM_001136538.1 | NP_001130010.1 | ||
| NM_025247.5 | NP_079523.3 | |||
| AK092356.1 | BAC03869.1 | |||
| AK310923.1 | ||||
| AL832043.1 | ||||
| AY323912.1 | AAQ88260.1 | |||
| BC015056.1 | AAH15056.1 | |||
| BC126358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630762 | Chr.12:111679417 - 111731317 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map