Assay Details
Target Gene Details
Entrez Gene ID: | 2641 |
Gene Name: | glucagon |
Gene Aliases: |
GLP1, GLP2, GRPP |
Location: |
Chr.2:162142869-162152404 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCG | NM_002054.4 | 6 | 857 | NP_002045.1 |
| AI818268.1 | 1 | 433 | ||
| BC005278.1 | 6 | 700 | AAH05278.1 | |
| BG654603.1 | 1 | 442 | ||
| CB069629.1 | 1 | 438 | ||
| J04040.1 | 6 | 638 | AAA52567.1 |
Target Gene Details
Entrez Gene ID: | 101929532 |
Gene Name: | uncharacterized LOC101929532 |
Gene Aliases: |
- |
Location: |
Chr.2:162114441-162172739 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929532 | NR_110255.1 | |||
| BX648358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834439 | Chr.2:162057118 - 162223456 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map