Assay Details
Target Gene Details
Entrez Gene ID: | 1016 |
Gene Name: | cadherin 18 |
Gene Aliases: |
CDH14, CDH14L, CDH24 |
Location: |
Chr.5:19472930-20575902 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1031377 | Chr.5:19289979 - 20971814 on Build GRCh38 | Loss |
|
| nsv597424 | Chr.5:19381946 - 19751051 on Build GRCh38 | Gain |
|
| nsv470997 | Chr.5:19441626 - 19534696 on Build GRCh38 | Loss |
|
| nsv4752 | Chr.5:19449761 - 19482880 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map