Assay Details
Target Gene Details
Entrez Gene ID: | 5143 |
Gene Name: | phosphodiesterase 4C |
Gene Aliases: |
DPDE1 |
Location: |
Chr.19:18207961-18248200 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE4C | NM_000923.4 | 16 | 2702 | NP_000914.2 |
| NM_001098818.2 | 15 | 2427 | NP_001092288.1 | |
| NM_001098819.2 | 15 | 2480 | NP_001092289.1 | |
| NR_040546.1 | 14 | 2377 | ||
| XM_011528056.2 | 15 | 2433 | XP_011526358.1 | |
| XM_011528058.2 | 13 | 2009 | XP_011526360.1 | |
| XM_017026866.1 | 16 | 2711 | XP_016882355.1 | |
| DB313915.1 | 1 | 406 | ||
| L20968.1 | AAA03591.1 | |||
| U66346.1 | AAB96875.1 | |||
| U66347.1 | AAB96876.1 | |||
| Z46632.1 | 15 | 2574 | CAA86601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
| nsv833775 | Chr.19:18102951 - 18279264 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map