Assay Details
Target Gene Details
Entrez Gene ID: | 79827 |
Gene Name: | CXADR like membrane protein |
Gene Aliases: |
ACAM, ASAM, CSBM, CSBS |
Location: |
Chr.11:123072006-123195305 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLMP | NM_024769.3 | 7 | 2201 | NP_079045.1 |
| AK026068.1 | 7 | 2177 | BAB15347.1 | |
| AY326422.1 | 7 | 2025 | AAP88386.1 | |
| AY358340.1 | 7 | 2025 | AAQ88706.1 | |
| BC009371.2 | 7 | 1847 | AAH09371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039284 | Chr.11:122985952 - 123083303 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map