Assay Details
Target Gene Details
Entrez Gene ID: | 8428 |
Gene Name: | serine/threonine kinase 24 |
Gene Aliases: |
HEL-S-95, MST3, MST3B, STE20, STK3 |
Location: |
Chr.13:98450199-98577151 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK24 | NM_001032296.3 | 11 | 3497 | NP_001027467.2 |
| NM_001286649.1 | 10 | 3440 | NP_001273578.1 | |
| NM_003576.4 | 11 | 3441 | NP_003567.2 | |
| EU668350.1 | 11 | 3488 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv562868 | Chr.13:98303535 - 98556312 on Build GRCh38 | Gain |
|
| nsv832695 | Chr.13:98387786 - 98582538 on Build GRCh38 | Loss |
|
| nsv471172 | Chr.13:98303135 - 98531805 on Build GRCh38 | Gain |
|
| nsv509511 | Chr.13:98413939 - 98492463 on Build GRCh38 | Insertion |
|
| nsv517409 | Chr.13:98436902 - 98463263 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map