Assay Details
Target Gene Details
Entrez Gene ID: | 6035 |
Gene Name: | ribonuclease A family member 1, pancreatic |
Gene Aliases: |
RAC1, RIB1, RNS1 |
Location: |
Chr.14:20801356-20802877 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE1 | NM_002933.4 | NP_002924.1 | ||
| NM_198232.2 | NP_937875.1 | |||
| NM_198234.2 | NP_937877.1 | |||
| NM_198235.2 | NP_937878.1 | |||
| AK098375.1 | ||||
| AK129871.1 | ||||
| AK312100.1 | ||||
| AV710596.1 | ||||
| BC005324.1 | AAH05324.1 | |||
| BC022882.1 | AAH22882.1 | |||
| BG539567.1 | ||||
| BI599530.1 | ||||
| BI753463.1 | ||||
| BI760598.1 | ||||
| BM007311.1 | ||||
| BM919747.1 | ||||
| CB993343.1 | ||||
| CD703706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428293 | Chr.14:20747949 - 21063624 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map