Assay Details
Target Gene Details
Entrez Gene ID: | 124152 |
Gene Name: | IQ motif containing K |
Gene Aliases: |
- |
Location: |
Chr.16:19716456-19858467 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCK | NM_001305121.1 | NP_001292050.1 | ||
| NM_001305122.1 | NP_001292051.1 | |||
| NM_153208.2 | NP_694940.1 | |||
| NR_130967.1 | ||||
| NR_130968.1 | ||||
| XM_011545736.2 | XP_011544038.1 | |||
| XM_017022933.1 | XP_016878422.1 | |||
| AF520569.1 | AAM77339.1 | |||
| AF520570.1 | AAM77340.1 | |||
| AK000122.1 | ||||
| AK093894.1 | ||||
| AK301933.1 | ||||
| AK303998.1 | ||||
| AK311216.1 | ||||
| AK315672.1 | ||||
| BC034823.1 | AAH34823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571582 | Chr.16:19038056 - 19790801 on Build GRCh38 | Gain |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
| nsv1037002 | Chr.16:19732327 - 19794287 on Build GRCh38 | Gain |
|

Genomic Map