Assay Details
Target Gene Details
Entrez Gene ID: | 88745 |
Gene Name: | ribosomal RNA processing 36 |
Gene Aliases: |
C6orf153, dJ20C7.4 |
Location: |
Chr.6:43021647-43029599 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRP36 | NM_033112.2 | NP_149103.1 | ||
| AF161371.1 | AAF28931.1 | |||
| AK313816.1 | ||||
| BC006293.2 | AAH06293.2 | |||
| BC011933.2 | AAH11933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462927 | Chr.6:42948182 - 43110718 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
| nsv462930 | Chr.6:43002769 - 43110718 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74538867] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map