Assay Details
Target Gene Details
Entrez Gene ID: | 79152 |
Gene Name: | fatty acid 2-hydroxylase |
Gene Aliases: |
FAAH, FAH1, FAXDC1, SCS7, SPG35 |
Location: |
Chr.16:74712955-74774831 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FA2H | NM_024306.4 | 7 | 1266 | NP_077282.3 |
| XM_011523319.2 | 7 | 965 | XP_011521621.1 | |
| AJ278219.1 | 6 | 914 | CAC20436.1 | |
| AK058016.1 | 7 | 1260 | BAB71632.1 | |
| AK303878.1 | 5 | 1018 | ||
| BC002679.2 | 7 | 1232 | AAH02679.2 | |
| BC004263.1 | 7 | 1232 | AAH04263.2 | |
| BC017049.1 | 7 | 1237 | AAH17049.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517513 | Chr.16:74709838 - 74778778 on Build GRCh38 | Loss |
|
| nsv952054 | Chr.16:74710903 - 74740502 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map