Assay Details
Target Gene Details
Entrez Gene ID: | 85464 |
Gene Name: | slingshot protein phosphatase 2 |
Gene Aliases: |
SSH-2, SSH-2L |
Location: |
Chr.17:29625938-29930239 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSH2 | NM_001282129.1 | 16 | 5958 | NP_001269058.1 |
| NM_033389.3 | 15 | 5689 | NP_203747.2 | |
| XM_005258058.3 | 17 | 5851 | XP_005258115.1 | |
| XM_005258059.2 | 14 | 5825 | XP_005258116.1 | |
| XM_005258060.3 | 3 | 4418 | XP_005258117.1 | |
| XM_006722149.3 | 14 | 9456 | XP_006722212.1 | |
| XM_011525402.2 | 16 | 5686 | XP_011523704.1 | |
| XM_011525404.2 | 14 | 6449 | XP_011523706.1 | |
| XM_011525405.2 | 14 | 6497 | XP_011523707.1 | |
| XM_011525406.2 | 13 | 5776 | XP_011523708.1 | |
| XM_011525407.1 | 13 | 5521 | XP_011523709.1 | |
| XM_017025251.1 | 16 | 5894 | XP_016880740.1 | |
| XM_017025252.1 | 14 | 5755 | XP_016880741.1 | |
| AB051512.1 | 3 | 4507 | BAB21816.1 | |
| AB099290.1 | 15 | 5649 | BAC97813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833410 | Chr.17:29607083 - 29776736 on Build GRCh38 | Loss |
|
| nsv833409 | Chr.17:29465646 - 29655807 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map