Assay Details
Target Gene Details
Entrez Gene ID: | 79718 |
Gene Name: | transducin (beta)-like 1 X-linked receptor 1 |
Gene Aliases: |
C21, DC42, IRA1, MRD41, TBLR1 |
Location: |
Chr.3:177019355-177197357 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBL1XR1 | NM_001321193.1 | NP_001308122.1 | ||
| NM_001321194.1 | NP_001308123.1 | |||
| NM_001321195.1 | NP_001308124.1 | |||
| NM_024665.5 | NP_078941.2 | |||
| XM_005247775.2 | XP_005247832.1 | |||
| XM_006713746.1 | XP_006713809.1 | |||
| XM_011513141.1 | XP_011511443.1 | |||
| XM_011513142.2 | XP_011511444.1 | |||
| XM_011513143.2 | XP_011511445.1 | |||
| XM_017007185.1 | XP_016862674.1 | |||
| AF268193.1 | AAG44736.1 | |||
| AF314544.1 | AAK00301.1 | |||
| AK022956.1 | BAB14331.1 | |||
| AK296946.1 | ||||
| BC113421.1 | ||||
| BX648935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518453 | Chr.3:177002992 - 177192806 on Build GRCh38 | Gain |
|
| esv3598783 | Chr.3:176984480 - 177075812 on Build GRCh38 | Gain |
|
| nsv1002405 | Chr.3:176723959 - 177453575 on Build GRCh38 | Gain |
|
| dgv4996n100 | Chr.3:176940347 - 177167118 on Build GRCh38 | Gain |
|
| esv3598784 | Chr.3:177012515 - 177159025 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map