Assay Details
Target Gene Details
Entrez Gene ID: | 51292 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
GMPR 2 |
Location: |
Chr.14:24232417-24239242 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
More Information
Set Membership: |
|

Genomic Map