Assay Details
Target Gene Details
Entrez Gene ID: | 100505385 |
Gene Name: | IQCJ-SCHIP1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.3:159069252-159897366 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCJ-SCHIP1 | NM_001197113.1 | NP_001184042.1 | ||
| NM_001197114.1 | NP_001184043.1 | |||
| AK096479.1 | ||||
| DQ157847.1 | ||||
| DQ157848.1 |
Target Gene Details
Entrez Gene ID: | 29970 |
Gene Name: | schwannomin interacting protein 1 |
Gene Aliases: |
SCHIP-1 |
Location: |
Chr.3:159273247-159897366 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCHIP1 | NM_001197107.1 | 1 | 598 | NP_001184036.1 |
| NM_001197108.1 | 1 | 598 | NP_001184037.1 | |
| NM_014575.3 | 1 | 598 | NP_055390.1 | |
| AF145713.1 | 1 | 93 | AAF34241.1 | |
| AF145714.1 | 1 | 93 | AAF34242.1 | |
| AF145715.1 | 1 | 93 | AAF34243.1 | |
| AK314662.1 | 1 | 541 | ||
| BC036535.2 | 1 | 601 | AAH36535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470971 | Chr.3:158873139 - 159440428 on Build GRCh38 | Gain |
|
| nsv998949 | Chr.3:159057017 - 159676761 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map