Assay Details
Target Gene Details
Entrez Gene ID: | 1981 |
Gene Name: | eukaryotic translation initiation factor 4 gamma 1 |
Gene Aliases: |
EIF-4G1, EIF4F, EIF4G, EIF4GI, P220, PARK18 |
Location: |
Chr.3:184314495-184335358 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4G1 | NM_001194946.1 | 11 | 1564 | NP_001181875.1 |
| NM_001194947.1 | 10 | 1507 | NP_001181876.1 | |
| NM_001291157.1 | 10 | 1596 | NP_001278086.1 | |
| NM_004953.4 | 3 | 1052 | NP_004944.3 | |
| NM_182917.4 | 9 | 1688 | NP_886553.3 | |
| NM_198241.2 | 10 | 1543 | NP_937884.1 | |
| NM_198242.2 | 6 | 1128 | NP_937885.1 | |
| NM_198244.2 | 7 | 1305 | NP_937887.1 | |
| AB210013.1 | 9 | 1682 | ||
| AF104913.1 | 7 | 1164 | AAC82471.1 | |
| AF281070.1 | 9 | 1546 | AAM69365.1 | |
| AK096719.1 | 8 | 1193 | ||
| AK128378.1 | 9 | 1292 | ||
| AK131407.1 | BAD18554.1 | |||
| AK226160.1 | 10 | 1477 | ||
| AK294567.1 | 8 | 1174 | ||
| AK299763.1 | 7 | 1233 | ||
| AK308319.1 | 5 | 893 | ||
| AL832754.1 | 6 | 1017 | ||
| AY082886.1 | 9 | 1546 | AAL92872.1 | |
| BC140892.1 | 11 | 1406 | ||
| BC140896.1 | 10 | 1349 | ||
| BX647812.1 | 10 | 1456 | CAI46013.1 | |
| D12686.1 | 3 | 1052 | BAA02185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422400 | Chr.3:184299495 - 184425713 on Build GRCh38 | Duplication |
|
| nsv829813 | Chr.3:184264997 - 184427996 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map