Assay Details
Target Gene Details
Entrez Gene ID: | 55929 |
Gene Name: | DNA methyltransferase 1 associated protein 1 |
Gene Aliases: |
DNMAP1, DNMTAP1, EAF2, MEAF2, SWC4 |
Location: |
Chr.1:44213428-44220679 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMAP1 | NM_001034023.1 | NP_001029195.1 | ||
| NM_001034024.1 | NP_001029196.1 | |||
| NM_019100.4 | NP_061973.1 | |||
| XM_005271037.4 | XP_005271094.1 | |||
| XM_005271039.1 | XP_005271096.1 | |||
| XM_005271040.1 | XP_005271097.1 | |||
| XM_006710771.3 | XP_006710834.1 | |||
| XM_011541784.2 | XP_011540086.1 | |||
| XM_011541785.2 | XP_011540087.1 | |||
| XM_017001807.1 | XP_016857296.1 | |||
| XM_017001808.1 | XP_016857297.1 | |||
| AB037846.1 | ||||
| AF265228.1 | AAF87079.1 | |||
| AK021605.1 | BAB13854.1 | |||
| AK054954.1 | ||||
| AK092898.1 | ||||
| AK096252.1 | ||||
| AK289366.1 | ||||
| AK293598.1 | ||||
| AK298793.1 | ||||
| AL136657.1 | CAB66592.1 | |||
| AL137200.1 | CAB69910.1 | |||
| BC002855.2 | AAH02855.1 | |||
| BC008053.2 | AAH08053.1 | |||
| BX537895.1 | CAD97886.1 |
More Information
Set Membership: |
|

Genomic Map