Assay Details
Target Gene Details
Entrez Gene ID: | 5550 |
Gene Name: | prolyl endopeptidase |
Gene Aliases: |
PE, PEP |
Location: |
Chr.6:105277567-105403124 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PREP | NM_002726.4 | 15 | 2408 | NP_002717.3 |
| AB020018.1 | 15 | 2175 | BAA86936.1 | |
| AB028867.1 | 15 | 2181 | BAB19053.1 | |
| BC030636.2 | 15 | 2368 | AAH30636.1 | |
| X74496.1 | 15 | 2175 | CAA52605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604438 | Chr.6:105256293 - 105421255 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map