Assay Details
Target Gene Details
Entrez Gene ID: | 7227 |
Gene Name: | transcriptional repressor GATA binding 1 |
Gene Aliases: |
GC79, LGCR |
Location: |
Chr.8:115408496-115669633 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPS1 | NM_001282902.2 | 6 | 9137 | NP_001269831.1 |
| NM_001282903.2 | 7 | 9626 | NP_001269832.1 | |
| NM_014112.4 | 7 | 9640 | NP_054831.2 | |
| XM_005251049.2 | 6 | 9455 | XP_005251106.1 | |
| XM_006716625.1 | 7 | 9577 | XP_006716688.1 | |
| XM_011517264.1 | 8 | 9710 | XP_011515566.1 | |
| XM_011517266.2 | 7 | 9445 | XP_011515568.1 | |
| XM_011517267.1 | 7 | 9563 | XP_011515569.1 | |
| XM_011517268.1 | 7 | 9552 | XP_011515570.1 | |
| XM_017013801.1 | 6 | 13239 | XP_016869290.1 | |
| AF183810.1 | 7 | 9634 | AAF23614.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428205 | Chr.8:115329343 - 115510682 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map