Assay Details
Target Gene Details
Entrez Gene ID: | 89910 |
Gene Name: | ubiquitin protein ligase E3B |
Gene Aliases: |
BPIDS, KOS |
Location: |
Chr.12:109477410-109543628 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE3B | NM_001270449.1 | 3 | 680 | NP_001257378.1 |
| NM_001270450.1 | 3 | 335 | NP_001257379.1 | |
| NM_001270451.1 | 3 | 347 | NP_001257380.1 | |
| NM_130466.3 | 3 | 680 | NP_569733.2 | |
| NM_183415.2 | 3 | 335 | NP_904324.1 | |
| XM_005253987.2 | 3 | 560 | XP_005254044.1 | |
| XM_006719681.3 | 3 | 893 | XP_006719744.1 | |
| XM_006719682.2 | 3 | 893 | XP_006719745.1 | |
| XM_011538959.2 | 2 | 442 | XP_011537261.1 | |
| XM_011538961.1 | 3 | 893 | XP_011537263.1 | |
| XM_017020195.1 | 3 | 548 | XP_016875684.1 | |
| XM_017020196.1 | 2 | 442 | XP_016875685.1 | |
| AF130072.1 | 1 | 72 | ||
| AF251046.2 | 3 | 650 | AAK28419.2 | |
| AK307385.1 | 2 | 214 | ||
| AL096740.1 | 3 | 649 | CAH56410.1 | |
| BC032301.1 | 3 | 313 | ||
| BC051266.1 | 3 | 651 | AAH51266.1 | |
| BC068221.1 | 3 | 317 | AAH68221.1 | |
| BC108705.1 | 3 | 311 | AAI08706.1 | |
| BC141880.1 | 3 | 558 | ||
| BC150637.1 | 3 | 311 | ||
| BX537403.1 | 3 | 325 | CAD97645.1 | |
| DA771082.1 | 3 | 335 |
More Information
Set Membership: |
|

Genomic Map