Assay Details
Target Gene Details
Entrez Gene ID: | 9179 |
Gene Name: | adaptor related protein complex 4 mu 1 subunit |
Gene Aliases: |
CPSQ3, MU-4, MU-ARP2, SPG50 |
Location: |
Chr.7:100101413-100107180 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4M1 | NM_004722.3 | 1 | 203 | NP_004713.2 |
| XM_005250689.4 | XP_005250746.1 | |||
| XM_005250690.4 | 1 | 203 | XP_005250747.1 | |
| XM_006716175.3 | 1 | 164 | XP_006716238.1 | |
| XM_011516685.2 | 1 | 164 | XP_011514987.1 | |
| XM_017012791.1 | XP_016868280.1 | |||
| AF020796.1 | 1 | 153 | AAD25869.1 | |
| AF155158.1 | 1 | 39 | AAD43328.1 | |
| AK304489.1 | ||||
| AK315782.1 | ||||
| BC018705.1 | 1 | 46 | AAH18705.1 | |
| DC413622.1 | 1 | 203 | ||
| Y08387.1 | 1 | 49 | CAA69667.1 |
Target Gene Details
Entrez Gene ID: | 4176 |
Gene Name: | minichromosome maintenance complex component 7 |
Gene Aliases: |
CDC47, MCM2, P1.1-MCM3, P1CDC47, P85MCM, PNAS146, PPP1R104 |
Location: |
Chr.7:100092728-100101940 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM7 | NM_001278595.1 | 1 | 232 | NP_001265524.1 |
| NM_005916.4 | 1 | 232 | NP_005907.3 | |
| AI083651.1 | 1 | 375 | ||
| DA397394.1 | 1 | 213 | ||
| DA538092.1 | 1 | 232 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| nsv951384 | Chr.7:100100678 - 100114877 on Build GRCh38 | Deletion |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map