Assay Details
Target Gene Details
Entrez Gene ID: | 26517 |
Gene Name: | translocase of inner mitochondrial membrane 13 |
Gene Aliases: |
TIM13, TIM13B, TIMM13A, TIMM13B, ppv1 |
Location: |
Chr.19:2425624-2427916 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIMM13 | NM_012458.3 | 3 | 1354 | NP_036590.1 |
| AK024764.1 | 3 | 989 | ||
| AL520160.3 | 1 | 603 | ||
| BQ015233.1 | 1 | 636 |
Target Gene Details
Entrez Gene ID: | 360200 |
Gene Name: | transmembrane protease, serine 9 |
Gene Aliases: |
- |
Location: |
Chr.19:2385254-2426261 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS9 | XM_011527978.2 | XP_011526280.1 | ||
| XM_011527980.1 | XP_011526282.1 | |||
| XM_011527982.2 | XP_011526284.1 | |||
| AK131261.1 | BAD18439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160562 | Chr.19:2328434 - 2460264 on Build GRCh38 | Deletion |
|
| nsv1064930 | Chr.19:2380093 - 2592252 on Build GRCh38 | Gain |
|
| nsv953944 | Chr.19:2283702 - 2525802 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map