Assay Details
Target Gene Details
Entrez Gene ID: | 6391 |
Gene Name: | succinate dehydrogenase complex subunit C |
Gene Aliases: |
CYB560, CYBL, PGL3, QPS1, SDH3 |
Location: |
Chr.1:161314376-161364751 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHC | NM_001035511.1 | 5 | 2159 | NP_001030588.1 |
| NM_001035512.1 | 5 | 2221 | NP_001030589.1 | |
| NM_001035513.1 | 4 | 2164 | NP_001030590.1 | |
| NM_001278172.1 | 4 | 2057 | NP_001265101.1 | |
| NM_003001.3 | 6 | 2323 | NP_002992.1 | |
| NR_103459.1 | 7 | 2350 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010776 | Chr.1:161312594 - 161439428 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115718047] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map