Assay Details
Target Gene Details
Entrez Gene ID: | 100529063 |
Gene Name: | BCL2L2-PABPN1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:23306762-23326185 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L2-PABPN1 | NM_001199864.1 | NP_001186793.1 |
Target Gene Details
Entrez Gene ID: | 8106 |
Gene Name: | poly(A) binding protein nuclear 1 |
Gene Aliases: |
OPMD, PAB2, PABII, PABP-2, PABP2 |
Location: |
Chr.14:23320188-23326185 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PABPN1 | NM_004643.3 | 1 | 890 | NP_004634.1 |
More Information
Set Membership: |
|

Genomic Map