Assay Details
Target Gene Details
Entrez Gene ID: | 153745 |
Gene Name: | family with sequence similarity 71 member B |
Gene Aliases: |
- |
Location: |
Chr.5:157162332-157166268 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71B | NM_130899.2 | 2 | 1127 | NP_570969.2 |
| AK057543.1 | 2 | 1123 | BAB71526.1 | |
| AY973323.1 | 2 | 1027 | ||
| BC022035.1 | 2 | 1135 | AAH22035.1 | |
| BC025397.1 | 2 | 1128 | AAH25397.1 | |
| BC025409.1 | 2 | 1128 | AAH25409.1 | |
| BC025998.1 | 2 | 1128 | AAH25998.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575892 | Chr.5:156975204 - 157220778 on Build GRCh38 | Gain |
|

Genomic Map