Assay Details
Target Gene Details
Entrez Gene ID: | 79148 |
Gene Name: | matrix metallopeptidase 28 |
Gene Aliases: |
EPILYSIN, MM28, MMP-25, MMP-28, MMP25 |
Location: |
Chr.17:35756249-35795707 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP28 | NM_024302.4 | 8 | 2139 | NP_077278.1 |
| NM_032950.3 | 9 | 2202 | NP_116568.1 | |
| NR_111988.1 | ||||
| XM_011525225.1 | 7 | 1914 | XP_011523527.1 | |
| XM_011525226.2 | 9 | 2251 | XP_011523528.1 | |
| XM_011525227.1 | 9 | 1880 | XP_011523529.1 | |
| XM_011525228.1 | 9 | 1905 | XP_011523530.1 | |
| XM_011525229.2 | 8 | 1780 | XP_011523531.1 | |
| XM_011525230.1 | 8 | 2005 | XP_011523532.1 | |
| XM_011525231.1 | XP_011523533.1 | |||
| XM_011525232.2 | 8 | 2061 | XP_011523534.2 | |
| XM_017025061.1 | 9 | 1869 | XP_016880550.1 | |
| XM_017025062.1 | 8 | 2135 | XP_016880551.1 | |
| XM_017025064.1 | XP_016880553.1 | |||
| AF330002.1 | 8 | 2084 | AAK01706.1 | |
| AK122604.1 | ||||
| BM702847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map