Assay Details
Target Gene Details
Entrez Gene ID: | 80174 |
Gene Name: | DBF4 zinc finger B |
Gene Aliases: |
ASKL1, CHIFB, DRF1, ZDBF1B |
Location: |
Chr.17:44708581-44752732 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBF4B | NM_025104.3 | NP_079380.2 | ||
| NM_145663.2 | NP_663696.1 | |||
| NR_036623.1 | ||||
| XM_006722098.2 | XP_006722161.1 | |||
| XM_006722102.3 | XP_006722165.1 | |||
| XM_011525281.2 | 5 | 1588 | XP_011523583.1 | |
| XM_017025145.1 | XP_016880634.1 | |||
| XM_017025146.1 | XP_016880635.1 | |||
| XM_017025147.1 | XP_016880636.1 | |||
| XM_017025148.1 | XP_016880637.1 | |||
| XM_017025149.1 | XP_016880638.1 | |||
| XM_017025150.1 | XP_016880639.1 | |||
| XM_017025151.1 | XP_016880640.1 | |||
| XM_017025152.1 | 5 | 1588 | XP_016880641.1 | |
| XM_017025153.1 | 5 | 1583 | XP_016880642.1 | |
| XM_017025154.1 | XP_016880643.1 | |||
| XM_017025155.1 | XP_016880644.1 | |||
| XM_017025156.1 | XP_016880645.1 | |||
| AF448801.1 | AAM73808.1 | |||
| AK023149.1 | 5 | 1672 | BAB14431.1 | |
| AK303282.1 | ||||
| BC016158.1 | AAH16158.1 | |||
| DA475446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833456 | Chr.17:44639788 - 44825873 on Build GRCh38 | Gain+Loss |
|
| nsv1067206 | Chr.17:44728495 - 44770726 on Build GRCh38 | Gain |
|
| esv3893016 | Chr.17:44723046 - 44776840 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map