Assay Details
Target Gene Details
Entrez Gene ID: | 4117 |
Gene Name: | male germ cell associated kinase |
Gene Aliases: |
RP62 |
Location: |
Chr.6:10762723-10838954 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAK | NM_001242385.1 | 13 | 2006 | NP_001229314.1 |
| NM_001242957.2 | 14 | 2127 | NP_001229886.1 | |
| NM_005906.5 | 13 | 2052 | NP_005897.1 | |
| NR_134935.1 | 15 | 2210 | ||
| NR_134936.1 | 15 | 2366 | ||
| XM_011514619.2 | 15 | 2266 | XP_011512921.1 | |
| XM_011514620.2 | 15 | 3053 | XP_011512922.1 | |
| XM_011514621.2 | 14 | 2249 | XP_011512923.1 | |
| XM_011514622.2 | 13 | 2383 | XP_011512924.1 | |
| XM_011514624.2 | 9 | 1222 | XP_011512926.1 | |
| XM_017010863.1 | 15 | 2579 | XP_016866352.1 | |
| XM_017010864.1 | 14 | 2502 | XP_016866353.1 | |
| XM_017010865.1 | 12 | 2054 | XP_016866354.1 | |
| AB593146.1 | 13 | 1960 | ||
| AF505623.1 | 13 | 2052 | AAN16405.1 | |
| AK314408.1 | 15 | 2363 | ||
| BC039825.1 | 14 | 2164 | AAH39825.1 | |
| JN226411.1 | 14 | 1898 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv966670 | Chr.6:10447438 - 10871656 on Build GRCh38 | Duplication |
|
| nsv600979 | Chr.6:10449025 - 10839526 on Build GRCh38 | Gain |
|
| nsv470800 | Chr.6:10538711 - 10863092 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map