Assay Details
Target Gene Details
Entrez Gene ID: | 56980 |
Gene Name: | PR domain 10 |
Gene Aliases: |
PFM7 |
Location: |
Chr.11:129899706-130003097 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDM10 | NM_020228.2 | 22 | 4207 | NP_064613.2 |
| NM_199437.1 | 21 | 4195 | NP_955469.1 | |
| NM_199438.1 | 18 | 3894 | NP_955470.1 | |
| NM_199439.1 | 16 | 3780 | NP_955471.1 | |
| XM_011542912.2 | 21 | 4418 | XP_011541214.1 | |
| XM_017018023.1 | 17 | 5967 | XP_016873512.1 | |
| AB033057.2 | 18 | 3678 | BAA86545.2 | |
| AF275817.1 | 18 | 3894 | AAF87243.1 | |
| AF503171.1 | 16 | 3711 | AAP30847.1 | |
| AL833172.1 | 21 | 4198 | ||
| BC112934.1 | 21 | 4187 | AAI12935.1 | |
| BX648944.1 | 22 | 4208 |
More Information
Set Membership: |
|

Genomic Map