Assay Details
Target Gene Details
Entrez Gene ID: | 283807 |
Gene Name: | F-box and leucine rich repeat protein 22 |
Gene Aliases: |
Fbl22 |
Location: |
Chr.15:63596225-63608898 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL22 | NM_203373.2 | 2 | 1048 | NP_976307.2 |
| XM_017022086.1 | XP_016877575.1 | |||
| BC065833.1 | 2 | 1004 | AAH65833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833034 | Chr.15:63500671 - 63657512 on Build GRCh38 | Loss |
|
| nsv827357 | Chr.15:63599565 - 63601982 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map