Assay Details
Target Gene Details
Entrez Gene ID: | 80344 |
Gene Name: | DDB1 and CUL4 associated factor 11 |
Gene Aliases: |
GL014, PRO2389, WDR23 |
Location: |
Chr.14:24114697-24125242 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF11 | NM_001163484.1 | NP_001156956.1 | ||
| NM_025230.4 | NP_079506.3 | |||
| NM_181357.2 | NP_852002.1 | |||
| NR_028099.1 | ||||
| NR_028100.1 | ||||
| AB209073.1 | BAD92310.1 | |||
| AF130070.1 | AAG35498.1 | |||
| AF267858.1 | AAG44727.1 | |||
| AK027223.1 | BAB15698.1 | |||
| AK057636.1 | ||||
| AK074296.1 | BAB85041.1 | |||
| AK091857.1 | ||||
| AK094478.1 | ||||
| AK292797.1 | ||||
| AK296742.1 | ||||
| AK307849.1 | ||||
| AL833831.1 | CAD38691.1 | |||
| BC008858.2 | AAH08858.2 | |||
| BC067132.1 | AAH67132.1 | |||
| BX161405.1 | CAD61885.1 | |||
| BX247970.1 | CAD62308.1 | |||
| BX648224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564057 | Chr.14:24056425 - 24149650 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map