Assay Details
Target Gene Details
Entrez Gene ID: | 3609 |
Gene Name: | interleukin enhancer binding factor 3 |
Gene Aliases: |
CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF110b, NF90, NF90a, NF90b, NFAR, NFAR-1, NFAR2, TCP110, TCP80 |
Location: |
Chr.19:10654261-10692419 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILF3 | NM_001137673.1 | 12 | 1638 | NP_001131145.1 |
| NM_004516.3 | 12 | 1638 | NP_004507.2 | |
| NM_012218.3 | 12 | 1638 | NP_036350.2 | |
| NM_017620.2 | 12 | 1638 | NP_060090.2 | |
| NM_153464.2 | 12 | 1638 | NP_703194.1 | |
| XM_006722742.2 | 12 | 1553 | XP_006722805.1 | |
| XM_011527984.2 | 12 | 1671 | XP_011526286.2 | |
| XM_011527985.2 | 12 | 1553 | XP_011526287.1 | |
| XM_011527987.1 | 8 | 932 | XP_011526289.1 | |
| XM_017026763.1 | 13 | 1762 | XP_016882252.1 | |
| XM_017026764.1 | 9 | 1226 | XP_016882253.1 | |
| XM_017026765.1 | 11 | 1455 | XP_016882254.1 | |
| AB209062.1 | 11 | 2258 | BAD92299.1 | |
| AF141870.1 | 12 | 1432 | AAD37575.1 | |
| AF147209.1 | 10 | 1333 | AAD33966.1 | |
| AF167569.1 | 10 | 1321 | AAD51098.1 | |
| AF167570.1 | 10 | 1321 | AAD51099.1 | |
| AJ271744.1 | 8 | 921 | CAC01404.1 | |
| AJ271745.1 | 11 | 1410 | CAC01405.1 | |
| AJ271746.1 | 11 | 1390 | CAC01406.1 | |
| AK291205.1 | 12 | 1537 | ||
| AK291617.1 | 12 | 1523 | ||
| AK293170.1 | 4 | 370 | ||
| AK294084.1 | 8 | 941 | ||
| BC064836.1 | 12 | 1571 | AAH64836.1 | |
| HQ322263.1 | 10 | 1321 | ||
| HQ322264.1 | 10 | 1321 | ||
| HQ322265.1 | 10 | 1321 | ||
| U10324.1 | 12 | 1588 | AAA20994.1 | |
| X98264.1 | CAA66917.1 | |||
| X98265.1 | 11 | 1530 | CAA66918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
| nsv1160585 | Chr.19:10674243 - 10771056 on Build GRCh38 | Deletion |
|
| nsv833746 | Chr.19:10678751 - 10842756 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map