Assay Details
Target Gene Details
Entrez Gene ID: | 4763 |
Gene Name: | neurofibromin 1 |
Gene Aliases: |
NFNS, VRNF, WSS |
Location: |
Chr.17:31094927-31377677 on Build GRCh38 |
Assay Gene Location: | Within Intron 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NF1 | NM_000267.3 | NP_000258.1 | ||
| NM_001042492.2 | NP_001035957.1 | |||
| XM_005257983.2 | XP_005258040.1 | |||
| XM_005257984.2 | XP_005258041.1 | |||
| XM_006721922.2 | XP_006721985.1 | |||
| XM_006721924.2 | XP_006721987.1 | |||
| XM_006721925.2 | XP_006721988.1 | |||
| XM_006721926.3 | XP_006721989.1 | |||
| XM_011524852.2 | XP_011523154.1 | |||
| XM_011524857.2 | XP_011523159.1 | |||
| XM_017024689.1 | XP_016880178.1 | |||
| AB209336.1 | BAD92573.1 | |||
| M38106.1 | AAA74897.1 | |||
| M38107.1 | AAB59558.1 | |||
| M82814.1 | AAA59924.1 | |||
| M89914.1 | AAA59925.1 |
Target Gene Details
Entrez Gene ID: | 4974 |
Gene Name: | oligodendrocyte myelin glycoprotein |
Gene Aliases: |
OMGP |
Location: |
Chr.17:31294650-31297362 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OMG | NM_002544.4 | NP_002535.3 | ||
| AK222663.1 | BAD96383.1 | |||
| AK312445.1 | ||||
| BC018050.1 | AAH18050.1 | |||
| BG704091.1 | ||||
| DA206834.1 | ||||
| M63623.1 | 1 | 1210 | AAA59970.1 | |
| X51694.1 | CAA35991.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833413 | Chr.17:31225070 - 31398610 on Build GRCh38 | Gain+Loss |
|
| nsv1061870 | Chr.17:31274278 - 31322138 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map