Assay Details
Target Gene Details
Entrez Gene ID: | 8667 |
Gene Name: | eukaryotic translation initiation factor 3 subunit H |
Gene Aliases: |
EIF3S3, eIF3-gamma, eIF3-p40 |
Location: |
Chr.8:116644816-116755823 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3H | NM_003756.2 | NP_003747.1 | ||
| AA902770.1 | ||||
| AK093128.1 | ||||
| AK222520.1 | BAD96240.1 | |||
| AK222620.1 | BAD96340.1 | |||
| AU117206.1 | ||||
| BC000386.2 | AAH00386.1 | |||
| BC090880.1 | AAH90880.1 | |||
| BT019594.1 | AAV38401.1 | |||
| CR456906.1 | CAG33187.1 | |||
| U54559.1 | AAD03465.1 |
More Information
Set Membership: |
|

Genomic Map