Assay Details
Target Gene Details
Entrez Gene ID: | 7351 |
Gene Name: | uncoupling protein 2 |
Gene Aliases: |
BMIQ4, SLC25A8, UCPH |
Location: |
Chr.11:73974671-73982844 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCP2 | NM_003355.2 | NP_003346.2 | ||
| AK025742.1 | ||||
| AK222540.1 | BAD96260.1 | |||
| AK222557.1 | BAD96277.1 | |||
| BC011737.2 | AAH11737.1 | |||
| U76367.1 | AAB48411.1 | |||
| U82819.1 | AAC51336.1 | |||
| U94592.1 | AAB53091.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520299 | Chr.11:73973249 - 73983709 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map