Assay Details
Target Gene Details
Entrez Gene ID: | 79023 |
Gene Name: | nucleoporin 37 |
Gene Aliases: |
p37 |
Location: |
Chr.12:102074189-102120139 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP37 | NM_024057.3 | 7 | 791 | NP_076962.2 |
| AF514994.1 | 6 | 645 | AAM76705.1 | |
| AK026271.1 | 6 | 626 | BAB15422.1 | |
| AK310619.1 | 7 | 751 | ||
| BC000861.2 | 6 | 709 | AAH00861.2 | |
| BM995521.1 | 4 | 520 |
More Information
Set Membership: |
|

Genomic Map