Assay Details
Target Gene Details
Entrez Gene ID: | 23428 |
Gene Name: | solute carrier family 7 member 8 |
Gene Aliases: |
LAT2, LPI-PC1 |
Location: |
Chr.14:23125295-23183660 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A8 | NM_001267036.1 | 9 | 2523 | NP_001253965.1 |
| NM_001267037.1 | 8 | 2268 | NP_001253966.1 | |
| NM_012244.3 | 11 | 3484 | NP_036376.2 | |
| NM_182728.2 | 9 | 2422 | NP_877392.1 | |
| NR_049767.1 | 7 | 2463 | ||
| AB037669.1 | 11 | 3447 | BAB21519.1 | |
| AF135830.1 | 7 | 2065 | AAF05697.1 | |
| AF171669.1 | 11 | 2961 | AAF20381.1 | |
| AK094550.1 | 3 | 1969 | ||
| AL365342.1 | 6 | 2816 | ||
| AL365344.1 | 5 | 1799 | ||
| AL365345.1 | 7 | 2101 | ||
| AL365346.1 | 3 | 1687 | ||
| AL365347.1 | 2 | 1075 | ||
| BC036825.1 | 7 | 2314 | AAH36825.1 | |
| BC052250.1 | 11 | 2767 | AAH52250.1 | |
| Y18483.1 | 11 | 3468 | CAB40137.1 |
More Information
Set Membership: |
|

Genomic Map