Assay Details
Target Gene Details
Entrez Gene ID: | 10848 |
Gene Name: | protein phosphatase 1 regulatory subunit 13 like |
Gene Aliases: |
IASPP, NKIP1, RAI, RAI4 |
Location: |
Chr.19:45379634-45406349 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R13L | NM_001142502.1 | 13 | 2718 | NP_001135974.1 |
| NM_006663.3 | 13 | 2716 | NP_006654.2 | |
| XM_017026177.1 | 14 | 2774 | XP_016881666.1 | |
| XM_017026178.1 | 14 | 2834 | XP_016881667.1 | |
| AF078036.1 | 7 | 1697 | AAD27004.1 | |
| AF078037.1 | 6 | 2151 | AAD27005.1 | |
| AY869712.1 | 13 | 2697 | AAW51146.1 | |
| BC001475.2 | 7 | 1303 | AAH01475.1 | |
| BC020589.1 | 6 | 957 | AAH20589.1 | |
| BC032298.1 | 13 | 2705 | AAH32298.1 | |
| BC064913.1 | 13 | 2681 | AAH64913.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833845 | Chr.19:45308814 - 45457371 on Build GRCh38 | Loss |
|
| esv3644502 | Chr.19:45323147 - 45403933 on Build GRCh38 | Gain |
|
| dgv1054e212 | Chr.19:45347119 - 45407663 on Build GRCh38 | Gain |
|
| nsv579867 | Chr.19:45323108 - 45408744 on Build GRCh38 | Gain |
|
| nsv1063547 | Chr.19:45322368 - 45400204 on Build GRCh38 | Gain |
|
| nsv833844 | Chr.19:45181051 - 45384148 on Build GRCh38 | Loss |
|
| dgv3606n100 | Chr.19:45347435 - 45400204 on Build GRCh38 | Gain |
|
| nsv442466 | Chr.19:45347447 - 45388666 on Build GRCh38 | Gain |
|
| nsv428365 | Chr.19:45358027 - 45469089 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map