Assay Details
Target Gene Details
Entrez Gene ID: | 9169 |
Gene Name: | SR-related CTD associated factor 11 |
Gene Aliases: |
CASP11, SFRS2IP, SIP1, SRRP129, SRSF2IP |
Location: |
Chr.12:45919131-45992040 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCAF11 | NM_004719.2 | 11 | 1559 | NP_004710.2 |
| XM_005269230.2 | 11 | 1449 | XP_005269287.2 | |
| XM_011538984.2 | 12 | 3078 | XP_011537286.1 | |
| XM_011538985.1 | 11 | 1393 | XP_011537287.1 | |
| XM_017020217.1 | 12 | 1437 | XP_016875706.1 | |
| XM_017020218.1 | 12 | 1437 | XP_016875707.1 | |
| XM_017020219.1 | 12 | 1636 | XP_016875708.1 | |
| XM_017020220.1 | 12 | 1437 | XP_016875709.1 | |
| XM_017020221.1 | 10 | 1766 | XP_016875710.1 | |
| XM_017020222.1 | 9 | 1736 | XP_016875711.1 | |
| XM_017020223.1 | 12 | 1659 | XP_016875712.1 | |
| AF030234.1 | 11 | 1507 | AAC39565.1 | |
| AK026473.1 | 12 | 1559 | ||
| AK302999.1 | 11 | 1484 | ||
| AK314093.1 | 5 | 822 | ||
| BC033589.1 | 7 | 964 | ||
| BC040951.2 | 3 | 640 | AAH40951.1 | |
| Y11251.1 | 1 | 1537 | CAA72121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832397 | Chr.12:45872207 - 46059950 on Build GRCh38 | Loss |
|
| esv2759897 | Chr.12:45755555 - 45956108 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map