Assay Details
Target Gene Details
Entrez Gene ID: | 8664 |
Gene Name: | eukaryotic translation initiation factor 3 subunit D |
Gene Aliases: |
EIF3S7, eIF3-p66, eIF3-zeta |
Location: |
Chr.22:36510850-36529431 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3D | NM_003753.3 | NP_003744.1 | ||
| AK300199.1 | ||||
| AK301284.1 | ||||
| AK303884.1 | ||||
| AK312939.1 | ||||
| BC000328.2 | AAH00328.1 | |||
| BC000469.2 | AAH00469.1 | |||
| BC014912.2 | AAH14912.1 | |||
| BC080515.1 | ||||
| BC093100.1 | AAH93100.1 | |||
| BC093686.1 | AAH93686.1 | |||
| BC101477.1 | AAI01478.1 | |||
| BT007381.1 | AAP36045.1 | |||
| CR456489.1 | CAG30375.1 | |||
| U54558.1 | AAD03466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760722 | Chr.22:36390921 - 36617980 on Build GRCh38 | Gain |
|
| esv2758836 | Chr.22:36510516 - 36645987 on Build GRCh38 | Loss |
|
| nsv520497 | Chr.22:36299896 - 36570195 on Build GRCh38 | Gain |
|
| nsv1060004 | Chr.22:36448479 - 36615303 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map