Assay Details
Target Gene Details
Entrez Gene ID: | 5734 |
Gene Name: | prostaglandin E receptor 4 |
Gene Aliases: |
EP4, EP4R |
Location: |
Chr.5:40679930-40740932 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGER4 | NM_000958.2 | 1 | 267 | NP_000949.1 |
| BC027934.1 | 1 | 267 | ||
| L25124.1 | 1 | 265 | AAA36438.1 | |
| L28175.1 | 1 | 63 | AAA36434.1 |
More Information
Set Membership: |
|

Genomic Map