Assay Details
Target Gene Details
Entrez Gene ID: | 5452 |
Gene Name: | POU class 2 homeobox 2 |
Gene Aliases: |
OCT2, OTF2, Oct-2 |
Location: |
Chr.19:42086110-42197931 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map