Assay Details
Target Gene Details
Entrez Gene ID: | 51768 |
Gene Name: | transmembrane 7 superfamily member 3 |
Gene Aliases: |
- |
Location: |
Chr.12:26966519-27014427 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM7SF3 | NM_016551.2 | 11 | 1596 | NP_057635.1 |
| XM_005253391.3 | 10 | 1464 | XP_005253448.1 | |
| XM_011520690.2 | 10 | 1459 | XP_011518992.1 | |
| XM_011520691.1 | XP_011518993.1 | |||
| XM_017019463.1 | 11 | 1707 | XP_016874952.1 | |
| XM_017019464.1 | 9 | 1269 | XP_016874953.1 | |
| XM_017019465.1 | 8 | 1116 | XP_016874954.1 | |
| XM_017019466.1 | 9 | 1283 | XP_016874955.1 | |
| AB032470.1 | 11 | 1395 | BAA92856.1 | |
| AK002031.1 | 11 | 1362 | BAA92046.1 | |
| AK023085.1 | 11 | 1584 | ||
| AK091115.1 | ||||
| BC005176.2 | 11 | 1521 | AAH05176.1 |
More Information
Set Membership: |
|

Genomic Map