Assay Details
Target Gene Details
Entrez Gene ID: | 8815 |
Gene Name: | barrier to autointegration factor 1 |
Gene Aliases: |
BAF, BCRP1, D14S1460, NGPS |
Location: |
Chr.11:66002079-66004149 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BANF1 | NM_001143985.1 | 1 | 284 | NP_001137457.1 |
| NM_003860.3 | 1 | 284 | NP_003851.1 | |
| AF068235.1 | 1 | 284 | AAD15901.1 | |
| BC107702.1 | AAI07703.1 | |||
| BG748852.1 | 1 | 67 | ||
| BU940708.1 | 1 | 101 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map