Assay Details
Target Gene Details
Entrez Gene ID: | 26509 |
Gene Name: | myoferlin |
Gene Aliases: |
FER1L3 |
Location: |
Chr.10:93306429-93482504 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 50 - Exon 51 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOF | NM_013451.3 | NP_038479.1 | ||
| NM_133337.2 | NP_579899.1 | |||
| XM_005269693.4 | XP_005269750.1 | |||
| XM_005269694.4 | XP_005269751.1 | |||
| XM_011539632.2 | XP_011537934.1 | |||
| XM_017016068.1 | XP_016871557.1 | |||
| XM_017016069.1 | XP_016871558.1 | |||
| XM_017016070.1 | XP_016871559.1 | |||
| AB033033.2 | BAA86521.2 | |||
| AF182316.1 | AAF27176.1 | |||
| AF207990.1 | AAG23737.1 | |||
| AK075258.1 | ||||
| AK093890.1 | ||||
| AL096713.1 | CAB46370.1 | |||
| BC052617.1 | AAH52617.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7494 | Chr.10:93316110 - 93351114 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map